Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arginine Hydrochloride, USP, 98.5-101.5%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
1 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
1119-34-2
C6H15ClN4O2
210.66
KWTQSFXGGICVPE-WCCKRBBISA-N
hydrogen (2S)-2-amino-5-[(diaminomethylidene)amino]pentanoic acid chloride
[H+].[Cl-].N[C@@H](CCCN=C(N)N)C(O)=O
1119-34-2
C6H15ClN4O2
210.66
KWTQSFXGGICVPE-WCCKRBBISA-N
hydrogen (2S)-2-amino-5-[(diaminomethylidene)amino]pentanoic acid chloride
[H+].[Cl-].N[C@@H](CCCN=C(N)N)C(O)=O

A1337, 1119-34-2, C6H14N4O2•HCl

Spectrum™ Arginine Hydrochloride, USP is a semiessential amino acid that has an important part in wound healing. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.002
Packaging Fiber Drum
Specific Rotation +21.4° to +23.6°
Sulfate 0.0003
Percent Purity 98.5 to 101.5%
Quantity 50 kg