Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Argininosuccinate Synthase Mouse anti-Human, Alexa Fluor™ 700, Clone: OTI1B10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270217AF700

Catalog No. N270217A700


Add to cart

Description

Description

Argininosuccinate Synthase Monoclonal antibody specifically detects Argininosuccinate Synthase in Human samples. It is validated for Western Blot.
Specifications

Specifications

Argininosuccinate Synthase
OTI1B10
Western Blot
Affinity Purified, Conjugated
ASS1
Synthetic peptide corresponding to residues near C-terminus of human Argininosuccinate Synthase (NP_000041).
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
argininosuccinate synthase, argininosuccinate synthase 1, argininosuccinate synthetase, argininosuccinate synthetase 1, ASSCTLN1, citrulline-aspartate ligase, Citrulline--aspartate ligase, EC 6.3.4.5
Mouse
IgG1
0.1 mL
Primary
445.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit