Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Argon (99.995%), Analytical Standard, MilliporeSigma™ Supelco™

Specifications:Contents: 14 liters Pressure: 240 psig Outlet fitting: CGA-160-1/8 inch, NPT F Weight: 1.5 lb.Dimensions: 3 x 11 inches D.O.T. specifications: 4B240

Manufacturer:  MilliporeSigma Supelco 501255

Catalog No. 11-100-1605


Add to cart

Description

Description

Specifications

Specifications

Gas
Analytical Standard
Cylinder
14 L
Safety and Handling

Safety and Handling

RTECSNumber : CF2300000

SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit