Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Argonaute 3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570415
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 92%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
AGO3 (protein argonaute-3) is required to RNA-mediated gene silencing (RNAi). It binds to short RNAs such as microRNAs (miRNAs) and represses the translation of mRNAs which are complementary to them. AGO3 is proposed to be involved in stabilization of small RNA derivates (riRNA) derived from processed RNA polymerase III-transcribed Alu repeats containing a DR2 retinoic acid response element (RARE) in stem cells and in the subsequent riRNA-dependent degradation of a subset of RNA polymerase II-transcribed coding mRNAs by recruiting a mRNA decapping complex involving EDC4.
Specifications
Argonaute 3 | |
Polyclonal | |
Unconjugated | |
AGO3 | |
AGO3; argonaute 3; argonaute 3, RISC catalytic component; argonaute RISC catalytic component 3; argonaute RISC catalytic subunit 3; argonaute3; AW048688; C130014L07Rik; eIF2C 3; eIF-2C 3; EIF2C1; EIF2C3; Eukaryotic translation initiation factor 2C 3; eukaryotic translation initiation factor 2C, 1; eukaryotic translation initiation factor 2C, 3; hAgo3; mAgo3; Piwi/Argonaute family protein meIF2C3; Protein argonaute-3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
214150 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8CJF9 | |
AGO3 | |
synthetic peptide directed towards the middle region of human Argonaut 3 (aa 160-209). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction