Learn More
Argos Technologies™ Accessories for Nova Microincinerator
Manufacturer:  Argos Technologies™ S3011H
 View more versions of this product
Catalog No. 03-391-192
Add to cart
Description
Description
Aluminum Three-Place Slide Dryer
  • Safely dries a variety of samples using the heat from your incinerator
  • Dries slides, sputums, exudates, and other thick mucous fluids
  • No morphological distortions
  • Easily attaches to the top of the burner face plate
  • Also dissolves epithelium using the KOH mycology technique
Aluminum Loop Rest
  • Protects loop handles from overheating or melting
  • Fits any loop
  • Attaches easily to all incinerators
Specifications
Specifications
UL, CE
1500°F
120V
Argos micro incinerator uses infrared heat to sterilize inoculating loops and needles
815°C