Learn More
Argos Technologies Accessories for Omega™ Pipet Controllers
Pipet with precision.  Accessories for Argos Technologies Omega™ Pipet Controllers are essential components for the most popular pipet aid with the colorful look and the weight balance.
Manufacturer:  Argos Technologies P5070
 View more versions of this product
Catalog No. 03-391-260
  Check Availability
Add to cart
Description
Description
  • Increased pipetting speed
  • Colorful look
  • Optimized weight balance
  • Ergonomic shape
  • Wide range of pipetting speed
  • Lithium-Polymer technology, which provides an extended cordless working time of over 6.5 hours non-stop
  • LED battery indicator clearly informs you about the charging status of the instrument
Specifications
Specifications
No
Omega™ pipet controller
Gray