Learn More
Argos Argos Technologies Black Microcentrifuge Tubes
Ideal for use with fluorescent probes or ethidium bromide
Manufacturer:  Argos T7456001
 View more versions of this product
Catalog No. 03-391-166
Add to cart
Description
Description
Ideal for use with fluorescent probes or ethidium bromide
Specifications
Specifications
Snap Cap
Yes
Black
15000xG
RNase, DNase-free
0.5mL
Polypropylene
Ideal for use with fluorescent probes or ethidium bromide