Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Argos Technologies™ E-Vac Aspirating Systems Filter

Argos Technologies E-Vac Aspirating Systems are a safe, compact, self-contained waste system providing an alternative to traditional in-house vacuum

Catalog No. 10-987-041

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

For use with Argos Technologies E-Vac Laboratory Aspiration System

  • Hydrophobic
  • Nonsterile
  • 60mm diameter

Specifications

Specifications

Hydrophobic membrane filter, non-sterile, 60mm diameter, 0.45 µm
6 cm
Hydrophobic PTFE
Hydrophobic Membrane Filter
2.36 in.
E-Vac™ laboratory aspiration system
1
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit