Learn More
Argos Technologies™ E-Vac Aspirating Systems
Argos Technologies E-Vac Aspirating Systems are a safe, compact, self-contained waste system providing an alternative to traditional in-house vacuum
Catalog No. 10-987-032
Add to cart
Includes: E-Vac 320 system includes 4L polypropylene bottle, lid with barbed tube fittings and HandEvac hand operator.
Description
Description
Ideal for use with critical or dangerous liquids or supernatants, pathogens, or in any class III or IV biohazard laboratories.
  • Quiet, oil-free membrane pump has a range from -250mm to 650mmHg
  • Once target vacuum is reached, pump automatically switches off
  • Pump automatically switches on when vacuum pressure is applied, allowing it to maintain a constant vacuum pressure
  • Perfect for gentle aspiration of small plates and dishes, as well as rapid emptying of larger containers
  • Easy-to-clean stainless-steel housing is UV-resistant for use in laminar hoods
  • E-Vac bottles can be autoclaved for 20 minutes at 121°C to prevent laboratory contamination
  • Base units are illuminated to allow visual inspection of waste levels in the bottle
  • Dual hydrophobic filters prevent liquids from reaching the pump housing and contaminating the system
  • Polypropylene bottles with lid fitted with standard barbed tube fittings or with quick-release tubing connectors for safe and convenient tubing connections (available separately)
  • Level detection sensor automatically shuts the pump off when the bottle is full
Certifications
CE approved, RoHS/WEEE compliant
Specifications
Specifications
-250 to -650 mbar
100/240 VAC
CE approved, RoHS/WEEE compliant
E-Vac 320 with 4L PP Polypropylene Bottle, lid with barbed tube fittings & HandEvac Handoperator
EV320
Universal Power Supply with US, UK, EU and SAA adapters
50/60 Hz
Analog dial
4L polypropylene Bottle, lid with barbed tube fittings & HandE-Vac hand operator
100/240 VAC 50/60 Hz
Glass
50 dB ± 5 dB at 1 m
8 L/min.
Oilless Diaphragm Pump