Learn More
Argos Technologies™ Flexirack Pipetting Workstation, Complete rack system with brackets
Versatile polypropylene rack functions as a pipetting workstation
$18.89
Specifications
Autoclavable No
View More Specs
Includes: Complete rack system (03-395-208) includes six 1.5mL tube inserts, two 0.5mL tube inserts, two 15mL tube inserts, two 50mL tube inserts, two racks, and two brackets.
Products 8
Catalog Number Type Color Price Quantity & Availability  
Catalog Number Type Color Price Quantity & Availability  
03-395-204
View Documents
Tube Insert Purple
Pack of 5 for $18.89
Add to cart
03-395-200
View Documents
V-Shape Basin Red This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
03-395-203
View Documents
Tube Insert Red This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
03-395-205
View Documents
Tube Insert Green This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
03-395-206
View Documents
Tube Insert Blue This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
03-395-207
View Documents
Tube Insert Orange This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
03-395-208
View Documents
Complete System with Brackets Rack Multi This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
03-395-201
View Documents
Rack without Brackets Multi This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
Versatile polypropylene rack functions as a pipetting workstation
  • Modular design allows you to create a rack based on your individual requirements
  • Can accommodate tubes from 0.5 to 50 mL depending upon which color-coded insert is used
  • Easy-release tabs allow user to quickly change inserts for multiple applications
  • Tiered rack design permits simultaneous viewing of all tubes
  • Small 10 × 5 × 3 in. (25.4 × 12.7 × 7.6cm) footprint and high-tube capacity maximize valuable laboratory space
Order Info
Rack can be purchased individually or as a set of two to double tube storage capacity. Disposable polystyrene basin inserts are also available.
Specifications
Specifications
No