Argos Technologies Flexirack Pipetting Workstation
Versatile polypropylene rack functions as a pipetting workstation
Manufacturer: Argos Technologies R6002
 View all options for this product
Catalog No. 03-395-202
  Check Availability
Add to Cart
    Autoclavable No
    For Use With (Equipment) FlexiRack pipetting workstation
    Quantity 6 /pack
    Product Type Brackets
    • Modular design allows you to create a rack based on your individual requirements
    • Can accommodate tubes from 0.5 to 50mL depending upon which color-coded insert is used
    • Easy-release tabs allow user to quickly change inserts for multiple applications
    • Tiered rack design permits simultaneous viewing of all tubes
    • Small 10 x 5 x 3 in. (25.4 x 12.7 x 7.6cm) footprint and high-tube capacity maximize valuable laboratory space
  • Description & Specifications
Related Products
Product Content Feedback