Learn More
Argos Technologies™ Nova Microcinerator
Burner assembly can be adjusted to five different positions for improved user comfort
Manufacturer:  Argos Technologies™ S3022
 View more versions of this product
Catalog No. 03-391-191
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Add to cart
Description
Description
Burner assembly can be adjusted to five different positions for improved user comfort
Certifications
UL listed, CSA approved, CE approved
Specifications
Specifications
UL, CSA, CE
1500°F
220V
Protects laboratory technicians from dangerous gases, flames and splatter
815°C