Learn More
Argos Technologies Nova Microcinerator
Burner assembly can be adjusted to five different positions for improved user comfort
Manufacturer:  Argos Technologies S3011
 View more versions of this product
Catalog No. 03-391-190
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
UL, CSA, CE
1500°F
120V
Protects laboratory technicians from dangerous gases, flames and splatter
815°C