Learn More
Argos Technologies RotoFlex™ Tube Rotator
Benchtop tube rotator designed for a variety of applications
Manufacturer:  Argos Technologies R2003
 View more versions of this product
Catalog No. 22-505-004
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
22rpm
RotoFlex and RotoFlex Plus benchtop tube rotator
1.5 to 2mL
CE, RoHS/WEEE
15 x 1.5 to 2mL Tubes
Tube rotisserie