Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARH Mouse anti-Human, Mouse, Rat, Canine, Alexa Fluor 488, Clone: OTI7A5, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271791AF488
Description
ARH Monoclonal specifically detects ARH in Human, Mouse, Rat, Canine samples. It is validated for Western Blot.Specifications
ARH | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
LDLRAP1 | |
Full length recombinant protein of human ARH (NP_056442) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine | |
IgG1 |
Western Blot | |
OTI7A5 | |
Western Blot | |
ARH1, ARH2, ARHAutosomal recessive hypercholesterolemia protein, DKFZp586D0624, FHCB1ARH1, FHCB2LDL receptor adaptor protein, low density lipoprotein receptor adapter protein 1, low density lipoprotein receptor adaptor protein 1, MGC34705 | |
Mouse | |
Affinity Purified | |
Lipid and Metabolism | |
26119.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title