Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARH Mouse anti-Human, Canine, Alexa Fluor™ 700, Clone: OTI7A5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271791AF700

Catalog No. N271791A700


Add to cart

Description

Description

ARH Monoclonal antibody specifically detects ARH in Human, Canine samples. It is validated for Western Blot.
Specifications

Specifications

ARH
OTI7A5
Western Blot
Affinity Purified, Conjugated
LDLRAP1
Full length recombinant protein of human ARH (NP_056442) produced in HEK293T cells.
Immunogen affinity purified
Lipid and Metabolism
Primary
26119.0
Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
ARH1, ARH2, ARHAutosomal recessive hypercholesterolemia protein, DKFZp586D0624, FHCB1ARH1, FHCB2LDL receptor adaptor protein, low density lipoprotein receptor adapter protein 1, low density lipoprotein receptor adaptor protein 1, MGC34705
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Canine, Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit