Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARHGAP19 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP19151220UL

 View more versions of this product

Catalog No. NBP1915220


Add to cart

Description

Description

ARHGAP19 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

ARHGAP19
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
ARHGAP19
Synthetic peptide directed towards the C terminal of human ARHGAP19. Peptide sequence: PTPESVAIGELKGTSKENRNLLFSGSPAVTMTPTRLKWSEGKKEGKKGFL
56 kDa
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Western Blot
Western Blot 1:1000
NP_116289
DKFZp313K217, MGC138804, MGC138805, MGC14258, putative RhoGAP protein, Rho GTPase activating protein 19, rho GTPase-activating protein 19, rho-type GTPase-activating protein 19
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
84986
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit