Learn More
Abnova™ ARHGAP24 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Human overexpression lysate
Manufacturer:  Abnova™ H00083478T01
Catalog No. 89-935-498
Add to cart
Description
Description
Specifications
Specifications
293T
NM_001042669
pCMV-ARHGAP24 full-length
100 μL
Human
ARHGAP24 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 73.3kDa
Transient overexpression cell lysate was tested with anti-ARHGAP24 antibody (Mfr. No. H00083478-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot