Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARHGAP25 Mouse anti-Human, Clone: 12G4, Abnova™

Mouse monoclonal antibody raised against full length recombinant human ARHGAP25.

Manufacturer:  Abnova Corporation MAB12855

Catalog No. 89-933-588


Add to cart

Description

Description

ARHGAPs, such as ARHGAP25, encode negative regulators of Rho GTPases (see ARHA; MIM 165390), which are implicated in actin remodeling, cell polarity, and cell migration (Katoh and Katoh, 2004 [PubMed 15254788]).[supplied by OMIM]

Specifications

Specifications

ARHGAP25
12G4
Mouse monoclonal antibody raised against full length recombinant human ARHGAP25.
In PBS, pH 7.3 (50% glycerol, 1% BSA, 0.02% sodium azide).
KAIA0053
Mouse
IgG1
50 uL
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Flow Cytometry (1:100) Immunofluorescence (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Liquid
ARHGAP25
Recombinant protein corresponding to full length human ARHGAP25.
Protein A/G purification
RUO
Primary
9938
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit