Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARHGAP25 Mouse anti-Human, Clone: OTI12G4, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA501745

Catalog No. 50-167-2862


Add to cart

Description

Description

ARHGAP25 encode negative regulators of Rho GTPases, which are implicated in actin remodeling, cell polarity, and cell migration.
Specifications

Specifications

ARHGAP25
Monoclonal
0.71 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P42331
HEL-S-308, KAIA0053
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
9938
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI12G4
Unconjugated
ARHGAP25
Liquid
ARHGAP25
Full length human recombit protein of human ARHGAP25 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit