Learn More
Aviva Systems Biology Corp ARHGAP28 Antibody - C-terminal region (ARP53789_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP53789_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ARHGAP28. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ARHGAP28. Alternate names: DKFZp686A2038, FLJ10312ApplicationWB. Species reactivity: Human
Catalog No. 50-014-61127