Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARHGAP28 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548983

Catalog No. PA548983

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

GTPase activator for the Rho-type GTPases by converting them to an inactive GDP-bound state.
Specifications

Specifications

ARHGAP28
Polyclonal
PBS with 0.09% sodium azide
Q9P2N2
Rho GTPase-activating protein 28, Rho-type GTPase-activating protein 28, ARHGAP28, KIAA1314
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79822
Western Blot
Unconjugated
ARHGAP28
Liquid
ARHGAP28
KLH conjugated synthetic peptide between 420-446 amino acids from the Central region of human ARHGAP28.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit