Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARHGAP36 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | ARHGAP36 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP174250
|
Novus Biologicals
NBP174250 |
100 μL |
Each of 1 for $436.00
|
|
Description
ARHGAP36 Polyclonal specifically detects ARHGAP36 in Human samples. It is validated for Western Blot.Specifications
ARHGAP36 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
FLJ30058, Rho GTPase activating protein 36, rho GTPase-activating protein 36 | |
ARHGAP36 | |
IgG | |
Affinity Purified |
Western Blot | |
Unconjugated | |
RUO | |
Q6ZRI8 | |
158763 | |
Synthetic peptides corresponding to the N terminal of RP13 102H20. 1. Immunizing peptide sequence KPDRALPIDRPNTLDKWFLILRGQQRAVSHKTFGISLEEVLVNEFTRRKH. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title