Learn More
Abnova™ ARHGAP9 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00064333T01
Catalog No. 89-935-518
Add to cart
Description
Description
Specifications
Specifications
293T
NM_032496.1
pCMV-ARHGAP9 full-length
100 μL
Human
ARHGAP9 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 82.61kDa
Transient overexpression cell lysate was tested with anti-ARHGAP9 antibody (Mfr. No. H00064333-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot