Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ARHGEF19 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569914
Description
This target displays homology in the following species: Cow: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%.
Guanine nucleotide exchange factors (GEFs) such as ARHGEF19 accelerate the GTPase activity of Rho GTPases (see RHOA, MIM 165390).[supplied by OMIM, Dec 2008].
Specifications
ARHGEF19 | |
Polyclonal | |
Unconjugated | |
ARHGEF19 | |
6030432F23; 6430573B13Rik; Arhgef19; Ephexin-2; Rho guanine nucleotide exchange factor (GEF) 19; Rho guanine nucleotide exchange factor 19; weakly similar to Rho GEF; weakly similar to Rho GEF 5; Wgef | |
Rabbit | |
Affinity chromatography | |
RUO | |
128272 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8IW93 | |
ARHGEF19 | |
synthetic peptide directed towards the middle region of human ARHGEF19. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction