Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ARHGEF37 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560542
Description
Immunogen sequence: KILENTVPDA SAYPVLQRAV SALQDVNTNI NEYKMRKEVA SKYTKVEQLT LRERLARINT HTLSKKTTRL Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 90%.
May act as a guanine nucleotide exchange factor (GEF).
Specifications
ARHGEF37 | |
Polyclonal | |
Unconjugated | |
ARHGEF37 | |
4933429F08Rik; ARHGEF37; probable guanine nucleotide exchange factor FLJ41603 homolog; protein 2-88; RGD1560471; Rho guanine nucleotide exchange factor (GEF) 37; Rho guanine nucleotide exchange factor 37; Scaffold protein tuba 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
389337 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A1IGU5 | |
ARHGEF37 | |
Recombinant protein corresponding to Human ARHGEF37. Recombinant protein control fragment (Product #RP-96970). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction