Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARHGEF39 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297431AF647
Description
ARHGEF39 Polyclonal specifically detects ARHGEF39 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ARHGEF39 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
C9orf100, chromosome 9 open reading frame 100, FLJ14642, MGC44886, Rho guanine nucleotide exchange factor (GEF) 39, RP11-331F9.7, vav-like protein C9orf100 | |
Produced in rabbits immunized with E. coli-derived Human ARHGEF39 fragment. | |
0.1 mL | |
Signal Transduction | |
84904 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction