Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARHGEF40 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 201101AP150UL
Specifications
ARHGEF40 | |
Polyclonal | |
Unconjugated | |
ARHGEF40 | |
ARHGEF40, FLJ10357, hypothetical protein FLJ10357, Protein SOLO, SOLO | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
268739, 55701 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3UPH7, Q8TER5 | |
ARHGEF40 | |
ARHGEF40 Fusion Protein Ag13886 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction