Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GEF5 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550117

Catalog No. PA550117


Add to cart

Description

Description

The antibody detects endogenous levels of total ARHGEF5 protein.

Rho GTPases play a fundamental role in numerous cellular processes initiated by extracellular stimuli that work through G protein coupled receptors. The encoded protein may form a complex with G proteins and stimulate Rho-dependent signals. This protein may be involved in the control of cytoskeletal organization.
Specifications

Specifications

GEF5
Polyclonal
Unconjugated
ARHGEF5
Liquid
ARHGEF5
Synthesized peptide derived from internal of human ARHGEF5.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q12774, E9Q7D5
ARH5, guanine nucleotide regulatory protein TIM, Oncogene TIM, p60 TIM, Rho guanine nucleotide exchange factor 5
Rabbit
IgG
100 μL
-20°C
Primary
54324, 7984
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit