Learn More
ARHGEF9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 200421AP
Catalog No. 50-173-2670
Add to cart
Description
Description
This antibody is specific to ARHGEF9.
The protein encoded by this gene is a Rho-like GTPase that switches between the active state and inactive state to regulate CDC42 and other genes. Defects in this gene are a cause of startle disease with epilepsy, also known as hyperekplexia with epilepsy. Three transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
ARHGEF9
0.39 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O43307, Q3UTH8, Q9QX73
ARHGEF9, COLLYBISTIN, HPEM 2, KIAA0424, PEM 2, PEM 2 homolog, PEM2
Rabbit
IgG
150 μL
-20°C
Primary
23229, 236915, 66013
Western Blot
Unconjugated
ARHGEF9
Liquid
Arhgef9
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat