Learn More
ARID1A Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 206521AP
Catalog No. 50-173-3014
Add to cart
Description
Description
The antibody is specific to ARID1A.
This gene encodes a member of the SWI/SNF family, whose members have helicase and ATPase activities and are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI, which is required for transcriptional activation of genes normally repressed by chromatin. It possesses at least two conserved domains that could be important for its function. First, it has a DNA-binding domain that can specifically bind an AT-rich DNA sequence known to be recognized by a SNF/SWI complex at the beta-globin locus. Second, the C-terminus of the protein can stimulate glucocorticoid receptor-dependent transcriptional activation. It is thought that the protein encoded by this gene confers specificity to the SNF/SWI complex and may recruit the complex to its targets through either protein-DNA or protein-protein interactions. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
ARID1A
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O14497
ARID1A, B120, BAF250, BAF250a, BM029, BRG1 associated factor 250, BRG1 associated factor 250a, C1orf4, hELD, hOSA1, Osa homolog 1, OSA1, P270, SMARCF1, SWI like protein, SWI/SNF complex protein p270
Rabbit
IgG
150 μL
-20°C
Primary
8289
Western Blot
Unconjugated
ARID1A
Liquid
ARID1A
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human