Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARID2 Rabbit anti-Human, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP303493100UL
Description
ARID2 Polyclonal antibody specifically detects ARID2 in Human samples. It is validated for Western Blot, KnockDownSpecifications
ARID2 | |
Unconjugated | |
Purified | |
Rabbit | |
IgG | |
100 μL | |
Store at -20°C. Avoid freeze-thaw cycles. | |
Polyclonal | |
Human |
KnockDown, Western Blot | |
PBS with 50% glycerol, pH7.3. | |
ARID domain-containing protein 2, AT rich interactive domain 2 (ARID, RFX-like), BAF200KIAA1557AT-rich interactive domain-containing protein 2, BRG1-associated factor 200, DKFZp686G052, DKFZp779P0222, FLJ30619, p200, Zinc finger protein with activation potential, zipzap, zipzap/p200 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1586-1835 of human ARID2 (NP_689854.2). | |
Affinity purified | |
RUO | |
Primary | |
196528 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title