Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARID2 Mouse anti-Human, Mouse, DyLight 650, Clone: 7311, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243567C

Catalog No. NBP243567C


Add to cart

Description

Description

ARID2 Monoclonal antibody specifically detects ARID2 in Human, Mouse samples. It is validated for Western Blot, Knockout Validated.
Specifications

Specifications

ARID2
7311
Western Blot, Knockout Validated
Purified
ARID2
Recombinant protein encompassing a sequence within the C-terminus region of human ARID2. The exact sequence is proprietary.
Protein G purified
Store at 4C in the dark.
Monoclonal
Human, Mouse
Western Blot, Immunoassay
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
ARID domain-containing protein 2, AT rich interactive domain 2 (ARID, RFX-like), BAF200KIAA1557AT-rich interactive domain-containing protein 2, BRG1-associated factor 200, DKFZp686G052, DKFZp779P0222, FLJ30619, p200, Zinc finger protein with activation potential, zipzap, zipzap/p200
Mouse
IgG1
0.1 ml
Primary
196528
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit