Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARID2 Rabbit anti-Human, Janelia Fluor 549, Polyclonal,Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP126615JF549

Catalog No. NBP12615549


Add to cart

Description

Description

ARID2 Polyclonal antibody specifically detects ARID2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

ARID2
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
ARID domain-containing protein 2, AT rich interactive domain 2 (ARID, RFX-like), BAF200KIAA1557AT-rich interactive domain-containing protein 2, BRG1-associated factor 200, DKFZp686G052, DKFZp779P0222, FLJ30619, p200, Zinc finger protein with activation potential, zipzap, zipzap/p200
Rabbit
IgG
0.1mL
Primary
196528
Immunohistochemistry, Immunohistochemistry (Paraffin)
Immunohistochemistry, Immunohistochemistry-Paraffin
Affinity Purified
ARID2
The immunogen recognized by this antibody maps to a region between residue 1785 and 1835 of human AT rich interactive domain 2 using the numbering given in entry NP_689854.2 (GeneID 196528).
Immunogen affinity purified
Store at 4°C in the dark.
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit