Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARID3C Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP17936320UL
Description
ARID3C Polyclonal specifically detects ARID3C in Human samples. It is validated for Western Blot.Specifications
ARID3C | |
Polyclonal | |
Western Blot 0.2-1 ug/ml | |
A6NKF2 | |
ARID3C | |
Synthetic peptide directed towards the C terminal of human ARID3CThe immunogen for this antibody is ARID3C (NP_001017363). Peptide Sequence: MGPMDPPRPCMPPSFLPRGKVPLREERLDGPLNLAGSGISSINMALEING | |
Affinity Purified | |
RUO | |
138715 | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
ARID domain-containing protein 3C, AT rich interactive domain 3C (BRIGHT- like), AT rich interactive domain 3C (BRIGHT-like), AT-rich interactive domain-containing protein 3C | |
Rabbit | |
44 kDa | |
20 μL | |
Primary | |
Human | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title