Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARID5A Mouse anti-Human, Mouse, DyLight 650, Clone: 18112, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243745C

Catalog No. NBP243745C


Add to cart

Description

Description

ARID5A Monoclonal antibody specifically detects ARID5A in Human, Mouse samples. It is validated for Western Blot.
Specifications

Specifications

ARID5A
18112
Western Blot
Purified
ARID5A
Recombinant protein encompassing a sequence within the center region of human ARID5A. The exact sequence is proprietary.
Protein A purified
Store at 4C in the dark.
Monoclonal
Human, Mouse
Western Blot
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
ARID domain-containing protein 5A, AT rich interactive domain 5A (MRF1-like), AT-rich interactive domain-containing protein 5A, RFVG5814
Mouse
IgG2b
0.1 ml
Primary
10865
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit