Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ARID5B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553270
Description
Immunogen sequence: TSKYPSRDMY RESENSSFPS HRHQEKLHVN YLTSLHLQDK KSAAAEAPTD DQPTDLSLPK NPHKPTGKVL GLAHSTTGPQ ESKGISQFQV LGSQSRDCHP KACRVSPMTM SGPKKYPESL SRSGKPHHVR LENFRKME Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 24%.
ARID5B encodes a member of the AT-rich interaction domain (ARID) family of DNA binding proteins. The encoded protein forms a histone H3K9Me2 demethylase complex with PHD finger protein 2 and regulates the transcription of target genes involved in adipogenesis and liver development. ARID5B also plays a role in cell growth and differentiation of B-lymphocyte progenitors, and single nucleotide polymorphisms in this gene are associated with acute lymphoblastic leukemia. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications
ARID5B | |
Polyclonal | |
Unconjugated | |
ARID5B | |
ARID domain-containing protein 5B; ARID5B; AT rich interactive domain 5B (MRF1-like); AT-rich interaction domain 5B; AT-rich interactive domain 5B (MRF1-like); AT-rich interactive domain-containing protein 5B; DESRT; Modulator recognition factor 2; modulator recognition factor 2 (MRF2); MRF1-like protein; MRF2; MRF-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84159 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q14865 | |
ARID5B | |
Recombinant protein corresponding to Human MRF2. Recombinant protein control fragment (Product #RP-90906). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction