Sartorius™ arium™ comfort I ASTM 1, 3 Combined Water purification Systems w/o UV Lamp, TOC Monitor

Use the Sartorius™ arium™ comfort I Combined System for producing ASTM Type 1 ultrapure water and Type 3 pure water.

Manufacturer: Sartorius™ H2OI2T

 View more versions of this product

Catalog No. 15-555-505

  Check Availability
Add to Cart

    Specifications

    Product Water Type Type 1 and Type 3
    Feedwater Pressure 2.0 - 6.9 bar
    Height (English) 19.7 in.
    Height (Metric) 50.1cm
    Width (English) 17 in.
    Width (Metric) 43.5cm
    Depth (English) 18.7 in.
    Depth (Metric) 47.6cm
    Daily Output 120L/hour (Type 1); 8L/hour or 16L/hour (Type 3)
    Turbidity <1NTU
    For Use With (Application) HPLC, GC-MS, AAS, ICP-MS, Ion chromatography, TOC-Analysis, Photometry, Buffer solutions, Feed water for laboratory devices, such as autoclaves, glassware washers etc
    Flow Rate 16L/hour
    Electrical Requirements 100-240VAC (+/-10%); 50-60Hz, 130VA (max.)
    Humidity Range 80%
    Feedwater Temperature (Metric) 2° - 30°C
    Includes UV lamp
    Particle Size <1mL
    Mounting Benchtop
    Purification Method Adsorption by spherical activated carbon, catalyst, reverse osmosis, deionization, UV radiation, optional final particle and sterile filtration
    Weight (English) 68.3 lb. (Operating); 50.7 lb. (Empty)
    Weight (Metric) Approx. 31kg (Operating); 23kg (Empty)
    Product Type Water purification System
    Bacterial Content <1 CFU/1,000mL

    • Unique cartridge kit for production of highest quality ultrapure water
    • Innovative iJust technology optimizes product water quality
    • Touch display with intuitive menu navigation
    • Compact design with SD card slot
    • Reproducible and reliable results
    • arium software controls valve on concentrate outlet corresponding to measurement data for CaCO3 and CO2
    • Available as bench-top system for every laboratory workstation and wall-mounted with integrated wall bracket
     
    Benefits
    • Optimized, economic water consumption
    • Premium product-water quality at all times
    • Guaranteed longer life of downstream ultrapure water systems

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit