Learn More
ARK5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 227231AP
Catalog No. 50-173-3934
Add to cart
Description
Description
Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. The STE group (homologs of yeast Sterile 7, 11, 20 kinases) consists of 50 kinases related to the mitogen-activated protein kinase (MAPK) cascade families (Ste7/MAP2K, Ste11/MAP3K, and Ste20/MAP4K). MAP kinase cascades, consisting of a MAPK and one or more upstream regulatory kinases (MAPKKs) have been best characterized in the yeast pheromone response pathway. Pheromones bind to Ste cell surface receptors and activate yeast MAPK pathway.
Specifications
Specifications
ARK5
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O60285, Q641K5
AMPK related protein kinase 5, ARK5, KIAA0537, NUAK family SNF1 like kinase 1, NUAK1
Rabbit
IgG
150 μL
-20°C
Primary
77976, 9891
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
NUAK1
Liquid
Nuak1
ARK5 Fusion Protein Ag18612
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse