Arkray™ DiaScreen™ Reagent Strips for Urinalysis
Manufacturer: ARKRAY USA INC D11000
 View all options for this product
Catalog No. 02-675-288
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Clia Complexity Waived
    Detectable Analytes Glucose, Protein, Ketone, pH, Blood, Bilirubin, Urobilinogen, Nitrite, Specific Gravity, Leukocyte
    For Use With (Application) Urinalysis, in-vitro diagnostic use
    For Use With (Equipment) Diascreen 50 Urine chemistry analyzer
    Quantity 100 Pack
    Test Time 60 or 120 sec.
    • Test strip spacing matches color chart, making strips easy to read
    • Only two read times for visual testing — 60 and 120 seconds
    • Dip-and-read test strips for in vitro diagnostic use
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback