Learn More
Arkray™ Dispette™ Tubes and Pipets
Manufacturer:  ACUGUARD CORP FH1500
 View more versions of this product
Catalog No. 13-664-38
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
CLSI
Each
Arkray™ Dispette™ pipet and reservoir
For 100 ESR determinations, for qualitative estimation of the erythrocyte sedimentation rate (ESR or sed-rate)
Room temperature