Arkray™ Micro Sed ESR System
Pipette tubes
Manufacturer: ACUGUARD CORP FH1530M
 View more versions of this product
Catalog No. 02-675-256
  Check Availability
Add to Cart
    Specifications
    Product Type Arkray™ Micro Sed™ micro pipette tube
    For Use With (Application) For pediatric use, qualitative estimation of the erythrocyte sedimentation rate, estimation of the ESR value on very small samples of Blood
    Quantity 250 Pack
    Storage Requirements Room temperature
    Certifications/Compliance CLSI, H02-A5
  • Description & Specifications
Related Products
Product Content Feedback