Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARL14EP Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297378PE
Description
ARL14EP Polyclonal antibody specifically detects ARL14EP in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
ARL14EP | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
ADP-ribosylation factor-like 14 effector protein, ARF7EP, C11orf46, chromosome 11 open reading frame 46, dJ299F11.1, FLJ38968, hypothetical protein LOC120534 | |
Produced in rabbits immunized with E. coli-derived Human ARL14EP fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
120534 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction