Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARL17A Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548244

Catalog No. PA548244

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

GTP-binding protein that functions as an allosteric activator of the cholera toxin catalytic subunit, an ADP- ribosyltransferase. Involved in protein trafficking; may modulate vesicle budding and uncoating within the Golgi apparatus.
Specifications

Specifications

ARL17A
Polyclonal
PBS with 0.09% sodium azide
Q8IVW1
ADP-ribosylation factor-like protein 17, ADP-ribosylation factor 7 variant, ARL17A, ARL17P1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51326
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ARL17A
Liquid
ARL17A
KLH conjugated synthetic peptide between 102-130 amino acids from the Central region of human ARL17P1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit