Learn More
ARL2BP Mouse anti-Human, Clone: 2G6, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA524446
Catalog No. PIMA524446
Add to cart
Description
Description
Peptide Sequence: MDALEGESFA LSFSSASDAE FDAVVGYLED IIMDDEFQLL QRNFMDKYYL EFEDTEENKL IYTPIFNEYI SLVEKYIEEQ LLQRIPEFNM AAFTTTLQHH KDEVAGDIFD MLLTFTDFLA FKEMFLDYRA EKEGRGLDLS SGLVVTSLCK SSSLPASQNN LRH
ADP-ribosylation factor (ARF)-like proteins (ARLs) comprise a functionally distinct group of the ARF family of RAS-related GTPases. The protein encoded by this gene binds to ARL2.GTP with high affinity but does not interact with ARL2.GDP. activated ARF.
Specifications
Specifications
ARL2BP
Monoclonal
Unconjugated
ARL2BP
Liquid
ARL2BP
ARL2BP (AAH03087, 1 a.a. ∽ 163 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Protein A
RUO
Antibody
Monoclonal
Human
ELISA, Western Blot
2G6
PBS with no preservative; pH 7.4
Q9Y2Y0
BART, BART1
Mouse
IgG1, kappa
100 μg
-20° C
Primary
23568