Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARL2BP Mouse anti-Human, PE/Cy5.5, Clone: OTI1A10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271743PECY55

Catalog No. N271743PEC5


Add to cart

Description

Description

ARL2BP Monoclonal antibody specifically detects ARL2BP in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

ARL2BP
OTI1A10
Flow Cytometry
Affinity Purified, Conjugated
ARL2BP
Full length recombinant protein of human ARL2BP (NP_036238) produced in E.coli.
Immunogen affinity purified
Signal Transduction
Primary
23568.0
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
ADP-ribosylation factor-like 2 binding protein, ADP-ribosylation factor-like protein 2-binding protein, ARF-like 2-binding protein, BART1binder of Arl Two, BARTArf-like 2 binding protein BART1, Binder of ARF2 protein 1, binder of Arl2
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit