Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARL3 Mouse anti-Human, Alexa Fluor™ 700, Clone: OTI1C6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270206AF700

Catalog No. N270206A700


Add to cart

Description

Description

ARL3 Monoclonal antibody specifically detects ARL3 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

ARL3
OTI1C6
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
ARL3
Recombinant protein fragment corresponding to amino acids 2-182 of human ARL3 (NP_004302) produced in E.coli.
Immunogen affinity purified
Signal Transduction
Primary
403.0
Immunohistochemistry, Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
ADP-ribosylation factor-like 3, ARFL3ADP-ribosylation factor-like protein 3, ARF-like 3
Mouse
IgG2a
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit