Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARL3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP169096
Description
ARL3 Polyclonal specifically detects ARL3 in Human samples. It is validated for Western Blot.Specifications
ARL3 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
ARL3 | |
Synthetic peptides corresponding to ARL3 (ADP-ribosylation factor-like 3) The peptide sequence was selected from the N terminal of ARL3. Peptide sequence QRKIRPYWKNYFENTDILIYVIDSADRKRFEETGQELAELLEEEKLSCVP. | |
Affinity purified | |
RUO | |
Primary | |
Yeast: 77%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Sheep, Yeast, Zebrafish | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
ADP-ribosylation factor-like 3, ARFL3ADP-ribosylation factor-like protein 3, ARF-like 3 | |
Rabbit | |
20 kDa | |
100 μL | |
Signal Transduction | |
403 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction