Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARL6IP4 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | ARL6IP4 |
---|---|
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
ARL6IP4 Polyclonal specifically detects ARL6IP4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ARL6IP4 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
51329 | |
IgG | |
Affinity purified |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
ADP-ribosylation-like factor 6 interacting protein 4, Aip-4, ARL-6-interacting protein 4, HSP-975, HSVI binding protein, HSVI-binding protein, MGC814, SFRS20, splicing factor, arginine/serine-rich 20, splicing regulator SRrp38, SR-15, SR-25ADP-ribosylation factor-like protein 6-interacting protein 4, SRp25 nuclear protein, SRp25aip-4, SRrp37 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 313-402 of human ARL6IP4 (NP_057722.3). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title