Learn More
ARL6IP6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 169561AP
Catalog No. 50-173-1292
Add to cart
Specifications
Specifications
ARL6IP6
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q68FV2, Q8BH07, Q8N6S5
Aip 6, ARL 6 interacting protein 6, ARL6IP6, PFAAP1
Rabbit
IgG
150 μL
-20°C
Primary
151188, 499798, 65103
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ARL6IP6
Liquid
Arl6ip6
ARL6IP6 Fusion Protein Ag10598
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat